Maleic acid ≥98%, Calbiochem®, Millipore®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

MX0100-20 MX0100-20
EM-MX0100-20EA 2923.19 USD
EM-MX0100-20 TXMX0100-20WRG
Maleic acid ≥98%, Calbiochem®, Millipore®
Maleic acid
Formula: C₄H₄O₄
Storage Temperature: Ambient
MDL Number: MFCD00063177
CAS Number: 110-16-7

Order Now

Specification Test Results

Assay (acidimetric) ≥ 99.0 %
Identity (IR) passes test
Melting range (lower value) ≥ 98 °C
Melting range (upper value) ≤ 103 °C
Spec. rotation α²0/D (c=5 in pyridine) -30.0 - -27.0 °


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR