Magnesium sulfate heptahydrate 98.0-102.0%, GR ACS, Supelco®

Supplier: MilliporeSigma

Synonyms: sulphuric acid magnesium salt heptahydrate, sulfuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

MX0070-20
EM-MX0070-3CS 1558.48 USD
EM-MX0070-3 EM-MX0070-20
Magnesium sulfate heptahydrate 98.0-102.0%, GR ACS, Supelco®
Magnesium sulphate heptahydrate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715

Order Now

Specification Test Results

Assay (complexometric) 98.0-102.0%
Ammonium (NH4) 0.002% max
Calcium (Ca) 0.02% max
Chloride (Cl) 5ppm max
Heavy metals (as Pb) 5ppm max
Insoluble matter 0.005% max
Iron (Fe) 5ppm max
Manganese (Mn) 5ppm max
Nitrate (NO3) 0.002% max
pH of a 5% solution at 25°C 5.0-8.2
Potassium (K) 0.005% max
Sodium (Na) 0.005% max
Strontium (Sr) 0.005% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR