Petroleum spirit, 60…90 °C, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

LX0260-3
EM-LX0260-3EA 290 USD
EM-LX0260-3
Petroleum spirit, 60…90 °C, Supelco®
Petroleum spirit

Practical

Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Boiling range 60-90°C
Color (APHA) 10 max
Density at 25°C 0.67-0.69g/mL
Form Clear liquid
Infrared spectrum Conforms to standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR