Iodomethane ≥98% stabilized, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Methyl iodide
IX0185-3
IX0185-1
EM-IX0185-3EA
1484
USD
EM-IX0185-3
EM-IX0185-1
Iodomethane ≥98% stabilized, Supelco®
Iodomethane
Stabilized with copper.
Formula:
CH₃I MW: 141.94 g/mol Boiling Pt: 42.4 °C (1013 hPa) Melting Pt: –66 °C Density: 2.279 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001073 CAS Number: 74-88-4 EINECS: 200-819-5 UN: 2644 ADR: 6.1,I Merck Index: 12,06161 |
Specification Test Results
Assay (GC) | 98% min |
Color (APHA) | 30 max |
Form | Clear liquid |
Infrared spectrum | Conforms to standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...