Iodine solution 0.1 N, Supelco®

Supplier: MilliporeSigma

IX0130A-4
EM-IX0130A-4CS 867 USD
EM-IX0130A-4
Iodine solution 0.1 N, Supelco®
Iodine

NIST Traceable.

Formula: I₂
MW: 253.81 g/mol
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3495
ADR: 8,III

Order Now

Specification Test Results

Expiration date Exp. 18 months
Normality 0.0995-0.1005N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR