Hydrobromic acid 48%, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hydrogen bromide

M-Clarity™ quality level = MQ200

HX0600-3
EM-HX0600-3EA 466 USD
EM-HX0600-3
Hydrobromic acid 48%, GR ACS, Supelco®
Hydrogen bromide

Meets ACS Specifications

Formula: HBr
MW: 80.91 g/mol
Boiling Pt: –66.7 °C (1013 hPa)
Melting Pt: –86 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1048
ADR: 2,
Merck Index: 13,04799

Order Now

Specification Test Results

Assay 47.0-49.0%
Chloride (Cl) 0.05% max
Color Yellow
Form Liquid
Heavy metals (as Pb) 5ppm max
Iodide (I) 0.003% max
Iron (Fe) 1ppm max
Phosphate (PO4) 0.001% max
Residue after ignition 0.002% max
Selenium (Se) 0.01ppm max
Sulfate and sulfite (as SO4) 0.003% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR