Hexane (mixture of isomers) ≥98.5%, OmniSolv® HR-GC for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

HX0297-1
EM-HX0297-1CS 1714 USD
EM-HX0297-1
Hexane (mixture of isomers) ≥98.5%, OmniSolv® HR-GC for gas chromatography, Supelco®
Hexane (mixture of isomers)

For high resolution gas chromatography. Purity based on n-hexane and saturated C6 hydrocarbons. Lot analysis and chromatograms on label.

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (n-hexane) Report
Assay (total aliphatic hydrocarbons C8) 99.9% min
Assay (total hexane isomers + methylcyclopentane) 98.5% min
Appearance Clear liquid, free from particulates
Cap. ECD responsive substances as PCNB (peak) 2ng/L max
Cap. ECD responsive substances as PCNB (total) 20ng/L max
Cap. FID responsive substances as decane (peak) 1µg/L max
Cap. FID responsive substances as decane (total) 10µg/L max
Color (APHA) 5 max
Filtered through 0.2 µm filter To pass test
Infrared spectrum Conforms
Residue after evaporation 1ppm max
Titrable acid 0.2µeq/g max
Water (H2O) 0.01% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR