n-Hexane ≥95%, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

HX0295-1 HX0295-6 HX0295-1 HX0295P-1
CAHX0295-1CS 3093.39 USD
CAHX0295-1 EM-HX0295-6 EM-HX0295-1 EM-HX0295P1
n-Hexane ≥95%, OmniSolv®, Supelco®
n-Hexane

Suitable for spectrophotometry, HPLC, and gas chromatography and residue analysis. UV cutoff 195nm. Certificate of Analysis on label.

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (n-hexane) 95% min
Boiling range 66.1-69.4°C
Capillary ECD responsive substances (as PCNB) 5ng/L max
Capillary FID responsive substances (as decane) 3µg/L max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 250ppt max
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 1ppm max
UV Abs. at 195 nm 1.00AU max
UV Abs. at 200 nm 0.50AU max
UV Abs. at 210 nm 0.20AU max
UV Abs. at 220 nm 0.05AU max
UV Abs. at 230 nm 0.05AU max
UV Abs. at 250 nm 0.01AU max
UV Abs. at 280 nm 0.005AU max
UV Cut-off 195nm max
Water (H2O) 0.01% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR