n-Heptane ≥99.0%, OmniSolv® for biosynthesis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: high purity solvents

HX0082-1
EM-HX0082-1EA 698 USD
EM-HX0082-1
n-Heptane ≥99.0%, OmniSolv® for biosynthesis, Supelco®
n-Heptane

Biosynthesis

Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.4 °C (1013 hPa)
Melting Pt: –90.6 °C
Density: 0.6838 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (n-heptane) 99.0% min
Assay (saturated C7 hydrocarbons) 99.9% min
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 1ppm max
UV Abs. at 200 nm 1.00AU max
UV Abs. at 210 nm 0.50AU max
UV Abs. at 220 nm 0.20AU max
UV Abs. at 250 nm 0.01AU max
UV Abs. at 270 nm 0.005AU max
UV Cut-off 200nm max
Water (H2O) 100ppm max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR