Formic acid ≥90%, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

FX0450-5
EM-FX0450-5CS 3124 USD
EM-FX0450-5
Formic acid ≥90%, Supelco®
Formic acid

Technical

Formula: HCO₂H
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 90% min
Color (APHA) 20 max
Form Clear liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR