Formic acid ≥88%, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Aminic Acid, Methanoic acid

M-Clarity™ quality level = MQ200

FX0445-6
EM-FX0445-6CS 3549 USD
EM-FX0445-6
Formic acid ≥88%, GR ACS, Supelco®
Formic acid

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: HCO₂H
MW: 46.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,
Merck Index: 13,04265
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acetic acid (CH3COOH) 0.4% max
Ammonium (NH4) 0.005% max
Assay 88.0% min
Chloride (Cl) 0.001% max
Color (APHA) 15 max
Dilution test To pass test
Evaporation residue 0.002% max
Heavy metals (as Pb) 5ppm max
Iron (Fe) 5ppm max
Sulfate (SO4) 0.002% max
Sulfite (SO3) To pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR