Water ASTM Type II, deionized distilled, Supelco®

Supplier: MilliporeSigma

EM3234-10
EM-EM3234-10EA 627 USD
EM-EM3234-10
Water ASTM Type II, deionized distilled, Supelco®
Water

Water, Deionized Distilled, ASTM Type II

Formula: H₂O
Boiling Pt: 100 °C (lit.)
Melting Pt: 0 °C (lit.)
Density: 1.000 g/ml at 3.98 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Chemical formula H₂O
Molar Mass 18.02
Storage RT
Chloride (Cl) 5ppb max
Conductivity at 25°C 1.0µS/cm max
Expiration date 18 months from mfg date
Form Clear liquid
Heavy metals (as Pb) 0.01ppm max
Iron (Fe) 0.3ppm max
Microbial limits 10 CFU/mL
Resistivity at 25°C 1.0MOhm·cm min
Silica (SiO2) 3ppb max
Sodium (Na) 5ppb max
Substances reducing permanganate To pass test
Total organic carbon 50µg/L max
Total solids 20ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR