1,4-Dioxane ≥99.9%, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

DX2091-6 DX2091-6
CADX2091-6EA 726.55 USD
CADX2091-6 EM-DX2091-1 EM-DX2091-6
1,4-Dioxane ≥99.9%, OmniSolv®, Supelco®
1,4-Dioxane

For spectrophotometry and chromatography. UV cutoff 215nm. Certificate of Analysis on label.

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101.1 °C (1013 hPa)
Melting Pt: 11.8 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3,II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Expiration date Exp 12 months from mfg date
Assay (GC) 99.9% min
Boiling range 101-102°C
Filtered through 0.2 µm filter To pass test
Peroxide (as H2O2) 2mg/L max
Refractive index (n 20/D) 1.4206-1.4226
Residue after evaporation 1mg/L max
UV Abs. at 215 nm 1.00AU max
UV Abs. at 250 nm 0.300AU max
UV Abs. at 300 nm 0.020AU max
UV Abs. at 350 nm 0.005AU max
UV Abs. at 400 nm 0.005AU max
UV Cut-off 215nm max
Water (H2O) 0.05% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR