N,N-Dimethylacetamide ≥99.9%, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

DX1543-1 DX1543-1
CADX1543-1EA 3221.38 USD
CADX1543-1 EM-DX1543-6 EM-DX1543-1
N,N-Dimethylacetamide ≥99.9%, OmniSolv®, Supelco®
N,N-Dimethylacetamide

For spectrophotometry and GC. UV cutoff 270nm. Certificate of Analysis on label.

Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 166.1 °C (1013 hPa)
Melting Pt: –20 °C
Density: 0.937 g/cm³ (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
UN: 2810
ADR: 6.1,III
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Color (APHA) 10 max
Density at 25°C 0.931-0.941g/mL
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 5ppm max
UV Abs. at 270 nm 1.00AU max
UV Abs. at 280 nm 0.30AU max
UV Abs. at 290 nm 0.15AU max
UV Abs. at 310 nm 0.05AU max
UV Abs. at 360 nm 0.01AU max
UV Cut-off 270nm max
Water (H2O) 0.05% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR