1,2-Dichloroethane, anhydrous ≥99.9%, DriSolv®, Supelco®

Supplier: MilliporeSigma
Danger


Packed in septum-sealed bottle.

DX0794-7
EM-DX0794-7CS 817 USD
EM-DX0794-7
1,2-Dichloroethane, anhydrous ≥99.9%, DriSolv®, Supelco®
1,2-Dichloroethane

Clear and free from particulates. Suitable for use in organic synthesis and other anhydrous solvent applications.
Elbow adapter with septum cap (EM-692004-2) allows for the removal of solvent under nitrogen atmosphere. Replacement septum (EM-692004S-2), for use with elbow adapter, minimizes air and moisture intrusion once the primary septum is punctured. Drying tube (EM-692010-2), filled with t.h.e.* desiccant indicating silica gel, removes solvent under a dry-air atmosphere. Large plug (EM-692012-2) is a replacement plug for bottle top.

Formula: ClH₂CCH₂Cl
Boiling Pt: 83.5 °C (1013 hPa)
Melting Pt: –35.5 °C
Density: 1.254 g/cm³ (20 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00000963
CAS Number: 107-06-2
EINECS: 203-458-1
UN: 1184
ADR: 3,II
Merck Index: 13,03831
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48789e88e6041156e33712fd7c0198d7a4910266&vid=4238623c54017660b83d3baa856a69cc1aa8ee6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.8% min
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Residue after evaporation 3ppm max
Water (H2O) 50ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR