1,2-Dichlorobenzene ≥98%, Supelco®

Supplier: MilliporeSigma
Warning

DX0685-3
EM-DX0685-3EA 816 USD
EM-DX0685-3
1,2-Dichlorobenzene ≥98%, Supelco®
1,2-Dichlorobenzene
Formula: C₆H₄Cl₂
Boiling Pt: 180.5 °C (1013 hPa)
Melting Pt: –17 °C
Density: 1.309 g/cm³ (20 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00000535
CAS Number: 95-50-1
EINECS: 202-425-9
UN: 1591
ADR: 6.1,III
Merck Index: 13,03081

Order Now

Specification Test Results

Assay (GC) 98% min
Color (APHA) 20 max
Form Clear liquid
Infrared spectrum Conforms to standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR