n-Decane ≥98%, Supelco®

Supplier: MilliporeSigma
Danger

DX0030-30
EM-DX0030-30EA 9645 USD
EM-DX0030-30
n-Decane ≥98%, Supelco®
n-Decane

Clear liquid. The infrared spectrum conforms to the standard. Partially soluble in water. Flammable and toxic effects.

Formula: H₃C(CH₂)₈CH₃
Boiling Pt: 174.1 °C (1013 hPa)
Melting Pt: –30 °C
Density: 0.73 g/cm³ (20 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00008954
CAS Number: 124-18-5
EINECS: 204-686-4
UN: 2247
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e4dead3a7841a2258e8a5b99b880895cd06aa1&vid=209e0ebbe6a38426575fcf5608cf6f2b586e6d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 98% min
Color (APHA) 10 max
Form Clear liquid
Infrared spectrum Conforms to standard
Melting Point –30°C (–22°F)
Vapor Density 4.9 kg/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR