Cyclopentane, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Pentamethylene

CX2414-1
EM-CX2414-1EA 693 USD
EM-CX2414-1
Cyclopentane, OmniSolv®, Supelco®
Cyclopentane

For spectrophotometry, liquid chromatography, and residue analysis. UV cutoff 198nm. Filtered through a 0.2µm element. Lot analysis on label.

Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 49
Melting Pt: -93.3
Density: 0.746 g/cm³ (20 °C)
Flash Pt: –41 °C
Storage Temperature: Ambient
MDL Number: MFCD00001356
CAS Number: 287-92-3
EINECS: 206-016-6
UN: 1146
ADR: 3 II
Merck Index: 13,02769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dd230057521cc77c90840c7adfece9c74c0a3b1&vid=352021851b7701a448c5b11d43e94d925d03f802&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (2,2-Dimethylbutane) 15.0% max
Assay (Cyclopentane) 85.0% min
Assay (n-Pentane) 3.0% max
Assay (Total) 99.8% min
Capillary ECD responsive substances (as PCNB) 5ng/L max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 500ppt max
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 1ppm max
UV Abs. at 198 nm 1.00AU max
UV Abs. at 210 nm 0.50AU max
UV Abs. at 220 nm 0.10AU max
UV Abs. at 230 nm 0.01AU max
UV Cut-off 198nm max
Water (H2O) 0.02% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR