Cyclohexanone ≥99% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Pimelic Ketone
CX2335-5
CX2335-7
CX2335-1
CX2335-20
CX2335-20
EM-CX2335-5EA
2903
USD
EM-CX2335-5
EM-CX2335-7
EM-CX2335-1
TXCX2335-20MNC
EM-CX2335-20
Cyclohexanone ≥99% (by GC), GR ACS ACS, Supelco®
Cyclohexanone
Meets ACS Specifications
Formula:
C₆H₁₀(=O) MW: 98.14 g/mol Boiling Pt: 155.6 Melting Pt: -31 Density: 0.95 g/cm3 (20 °C) Flash Pt: 43 Storage Temperature: Ambient |
MDL Number:
MFCD00001625 CAS Number: 108-94-1 EINECS: 203-631-1 UN: 1915 ADR: 3 III REACH: 01-2119453616-35 Merck Index: 13,02755 |
|
Specification Test Results
Expiration date | Exp 24 months from receipt date |
Assay (GC) | 99.0% min |
Color (APHA) at time of manufacture | 10 max |
Form | Clear liquid |
Infrared spectrum | Conforms to standard |
Residue after evaporation | 0.05% max |
Water (H2O) | 0.05% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...