Cyclohexane ≥99.0%, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hexahydro-Benzene, Hexahydrobenzene, Hexamethylene

CX2286-1 CX2286-1 CX2286-6
CACX2286-1CS 3882.68 USD
CACX2286-1 EM-CX2286-1 EM-CX2286-6
Cyclohexane ≥99.0%, OmniSolv®, Supelco®
Cyclohexane

For spectrophotometry, chromatography, and residue analysis. UV cutoff 200nm. Certificate of Analysis on label.

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81
Melting Pt: 7
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3 II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.0% min
Capillary ECD responsive substances (as PCNB) 5ng/L max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 250ppt max
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 1ppm max
UV Abs. at 200 nm 1.00AU max
UV Abs. at 210 nm 0.50AU max
UV Abs. at 220 nm 0.20AU max
UV Abs. at 230 nm 0.10AU max
UV Abs. at 240 nm 0.05AU max
UV Abs. at 250 nm 0.01AU max
UV Abs. at 260 nm 0.01AU max
UV Abs. at 270 nm 0.005AU max
UV Cut-off 200nm max
Water (H2O) 0.01% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR