Cyclohexane ≥99.0%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Hexahydro-Benzene, Hexahydrobenzene, Hexamethylene
CX2286-1
CX2286-1
CX2286-6
CACX2286-1CS
3882.68
USD
CACX2286-1
EM-CX2286-1
EM-CX2286-6
Cyclohexane ≥99.0%, OmniSolv®, Supelco®
Cyclohexane
For spectrophotometry, chromatography, and residue analysis. UV cutoff 200nm. Certificate of Analysis on label.
Formula:
C₆H₁₂ MW: 84.16 g/mol Boiling Pt: 81 Melting Pt: 7 Density: 0.7785 g/cm³ (20 °C) Flash Pt: –18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003814 CAS Number: 110-82-7 EINECS: 203-806-2 UN: 1145 ADR: 3 II Merck Index: 13,02752 |
|
Specification Test Results
Assay (GC) | 99.0% min |
Capillary ECD responsive substances (as PCNB) | 5ng/L max |
Color (APHA) | 10 max |
Filtered through 0.2 µm filter | To pass test |
Fluorescence (as quinine base) | 250ppt max |
Form | Clear liquid |
Infrared spectrum | Conforms to standard |
Residue after evaporation | 1ppm max |
UV Abs. at 200 nm | 1.00AU max |
UV Abs. at 210 nm | 0.50AU max |
UV Abs. at 220 nm | 0.20AU max |
UV Abs. at 230 nm | 0.10AU max |
UV Abs. at 240 nm | 0.05AU max |
UV Abs. at 250 nm | 0.01AU max |
UV Abs. at 260 nm | 0.01AU max |
UV Abs. at 270 nm | 0.005AU max |
UV Cut-off | 200nm max |
Water (H2O) | 0.01% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...