Citric acid, anhydrous 99.5-100.5% (alkalimetric, calculated on anhydrous substance), GR ACS, Supelco®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
CX1723-7
CX1723-20
CX1723-3
CX1723-1
EM-CX1723-7EA
1054
USD
EM-CX1723-7
EM-CX1723-20
EM-CX1723-3
EM-CX1723-1
Citric acid, anhydrous 99.5-100.5% (alkalimetric, calculated on anhydrous substance), GR ACS, Supelco®
Citric acid
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Formula:
HOC(COOH)(CH₂COOH)₂ MW: 192.13 g/mol Boiling Pt: 200 °C (1013 hPa) (dec.) Melting Pt: 153…159 °C (lit.) Density: 1.67 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011669 CAS Number: 77-92-9 EINECS: 201-069-1 REACH: 01-2119457026-42-0000 Merck Index: 13,02350 |
|
Specification Test Results
Assay (alkalimetric on dried substance) | 99.5-100.5% |
Chloride (Cl) | 0.001% max |
Residue after ignition | 0.02% max |
Insoluble matter | 0.005% max |
Iron (Fe) | 0.0003% max |
Lead (Pb) | 0.0002% max |
Oxalate (C2O4) |
To pass test ( |
Phosphate (PO4) | 0.001% max |
Substances carbonized by hot sulfuric acid | To pass test |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...