Chloroform ≥99.8% stabilised, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

CX1059-1 CX1059-1
CACX1059-1CS 2561.62 USD
CACX1059-1 EM-CX1059-6 EM-CX1059-1
Chloroform ≥99.8% stabilised, OmniSolv®, Supelco®
Chloroform

For spectrophotometry, HPLC, GC, and residue analysis. Stabilized with 1% ethanol (amylene free). Filtered through a 02µm element. UV cutoff 245nm. Certificate of Analysis on label.

Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7
Melting Pt: -63
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (CHCl3 + C2H5OH) 99.8% min
Capillary ECD responsive substances (as PCNB) 5ng/L max
Color (APHA) 10 max
Ethanol 0.8-1.2%
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 250ppt max
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 1ppm max
Titrable acid 0.2µeq/g max
UV Abs. at 245 nm 1.00AU max
UV Abs. at 255 nm 0.15AU max
UV Abs. at 260 nm 0.05AU max
UV Abs. at 270 nm 0.02AU max
UV Abs. at 290 nm 0.01AU max
UV Cut-off 245nm max
Water (H2O) 0.01% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR