Chlorobenzene ≥99.5% (by GC), GR ACS, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

CX0855-30
EM-CX0855-3CS 3794.86 USD
EM-CX0855-3 EM-CX0855-30
Chlorobenzene ≥99.5% (by GC), GR ACS, Supelco®
Chlorobenzene

Meets ACS Specifications

Formula: C₆H₅Cl
MW: 112.56 g/mol
Boiling Pt: 132 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.11 g/cm3 (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.5% min
Color (APHA) 30 max
Form Clear liquid
Residue after evaporation 20ppm max
Titrable acid 0.004meq/g max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR