Carbon disulfide ≥99.9%, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

CX0396-6
EM-CX0396-6CS 900 USD
EM-CX0396-6
Carbon disulfide ≥99.9%, OmniSolv®, Supelco®
Carbon disulphide

Meets Reagent Specifications for testing USP/NF monographs

Formula: CS₂
MW: 76.14 g/mol
Boiling Pt: 46.2 °C (1013 hPa)
Melting Pt: –111.6 °C
Density: 1.2632 g/cm³ (20 °C)
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,I
Merck Index: 13,01821
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 5ppm max
UV Abs. at 382 nm 1.00AU max
UV Abs. at 388 nm 0.50AU max
UV Abs. at 403 nm 0.10AU max
UV Abs. at 410 nm 0.05AU max
UV Abs. at 500 nm 0.005AU max
UV Cut-off 382nm max
Water (H2O) 0.03% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR