tert-Butanol ≥99% (by GC), GR ACS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
BX1805-1
BX1805-4
BX1805-2
BX1805-5
BX1805-6
EM-BX1805-1CS
1486
USD
EM-BX1805-1
EM-BX1805-4
EM-BX1805-2
EM-BX1805-5
EM-BX1805-6
tert-Butanol ≥99% (by GC), GR ACS, Supelco®
tert-Butanol
Meets ACS Specifications.
Formula:
(CH₃)₃COH Boiling Pt: 81…83 °C (1013 hPa) Melting Pt: 24…25 °C Density: 0.78 g/cm3 (20 °C) Flash Pt: 11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004464 CAS Number: 75-65-0 EINECS: 200-889-7 UN: 1120 ADR: 3,II Merck Index: 12,01577 |
|
Specification Test Results
Assay (GC) | 99.0% min |
Carbonyl compounds (as HCHO) | 0.01% max |
Color (APHA) | 20 max |
Form | Clear liquid (or solid, m.p. 24°C) |
Infrared spectrum | Conforms to standard |
Residue after evaporation | 0.003% max |
Titrable acid | 0.001meq/g max |
Water (H2O) | 0.1% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...