Isobutanol ≥99.0%, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

BX1791-5 BX1791-1
EM-BX1791-5EA 672 USD
EM-BX1791-5 EM-BX1791-1
Isobutanol ≥99.0%, GR ACS, Supelco®
Isobutanol

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: (CH₃)₂CHCH₂OH
Boiling Pt: 107.9 °C (1013 hPa)
Melting Pt: –108 °C
Density: 0.802 g/cm³ (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3,III
Merck Index: 13,05148
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7c83a107c7a71fa54ed2b1bbdfb78617fe3f67&vid=df3ce90fc7ae8dafb7fabe31b244825735325030&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.0% min
Carbonyl compounds (as 2-butanone) 0.02% max
Carbonyl compounds (as butyraldehyde) 0.01% max
Color (APHA) 10 max
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 0.001% max
Solubility in water To pass test
Titrable acid 0.0005meq/g max
Water (H2O) 0.1% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR