1-Butanol ≥99.4% (by GC), GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

BX1780-6 BX1780-20 BX1780-5
EM-BX1780-6CS 692 USD
EM-BX1780-6 EM-BX1780-20 EM-BX1780-5
1-Butanol ≥99.4% (by GC), GR ACS, Supelco®
1-Butanol

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: H₃C(CH₂)₂CH₂OH
Boiling Pt: 116…118 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.81 g/cm3 (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.4% min
Carbonyl compounds (as butyraldehyde) 0.01% max
Color (APHA) 10 max
Di-n-Butyl ether [(C4 H9)2 O] 0.2% max
Form Clear liquid
Residue after evaporation 0.005% max
Titrable acid 0.0008meq/g max
Water (H2O) 0.1% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR