Methyl ethyl ketone ≥99.5%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
EK 500 Additive, Butan-2-one, Ethyl methyl ketone, 2-Butanone, Butanone, MEK
BX1673-3
BX1673P-1
EM-BX1673-3CS
710
USD
EM-BX1673-3
EM-BX1673P-1
Methyl ethyl ketone ≥99.5%, OmniSolv®, Supelco®
Methyl ethyl ketone
For spectophotometry and chromatography. UV cutoff 329nm. Lot analysis on label.
Formula:
C₂H₅COCH₃ MW: 72.11 g/mol Boiling Pt: 79.6 °C (1013 hPa) Melting Pt: –86 °C Density: 0.8054 g/cm³ (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011648 CAS Number: 78-93-3 EINECS: 201-159-0 UN: 1193 ADR: 3,II REACH: 01-2119457290-43 Merck Index: 13,06097 |
Specification Test Results
Assay (GC) | 99.5% min |
Color (APHA) | 10 max |
Filtered through 0.2 µm filter | To pass test |
Form | Clear liquid |
Infrared spectrum | Conforms to standard |
Residue after evaporation | 5ppm max |
UV Abs. at 329 nm | 1.00AU max |
UV Abs. at 335 nm | 0.30AU max |
UV Abs. at 344 nm | 0.05AU max |
UV Abs. at 350 nm | 0.01AU max |
UV Abs. at 400 nm | 0.005AU max |
UV Cut-off | 329nm max |
Water (H2O) | 0.05% max |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...