Benzyl alcohol ≥99.0%, GR ACS, Supelco®

Supplier: MilliporeSigma
Warning

BX0483-3
EM-BX0483-3CS 3361 USD
EM-BX0483-3
Benzyl alcohol ≥99.0%, GR ACS, Supelco®
Benzyl alcohol

Meets ACS Specifications

Formula: C₆H₅CH₂OH
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –15.3 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.0% min
Acetophenone 0.02% max
Acidity 0.01meq/g max
Benzaldehyde 0.01% max
Chlorinated compounds To pass test
Color (APHA) 20 max
Form Clear liquid
Identification To pass test
Refractive index (n 20/D) 1.539-1.541
Residue after evaporation 0.05% max
Residue after ignition 0.005% max
Specific Gravity at 25/25°C 1.042-1.047


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR