Benzene ≥99% (by GC), GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

BX0220-5 BX0220-20 BX0220-6 BX0220-3 BX0220-5
CABX0220-5CS 3347.4 USD
CABX0220-5 EM-BX0220-20 EM-BX0220-6 EM-BX0220-3 EMD-BX0220-5
Benzene ≥99% (by GC), GR ACS, Supelco®
Benzene

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: C₆H₆
Boiling Pt: 80.1 °C (1013 hPa)
Melting Pt: 5.5 °C
Density: 0.88 g/cm3 (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00003009
CAS Number: 71-43-2
EINECS: 200-753-7
UN: 1114
ADR: 3,II
REACH: 01-2119447106-44
Merck Index: 13,01066
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2c0146712ab33142aeeb3843b02381e6bda4028&vid=cae270bfe08ae6ab4b14d9e1775b89a82cfeec1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.0% min
Color (APHA) 10 max
Form Clear liquid
Residue after evaporation 0.001% max
Substances darkened by sulfuric acid To pass test
Sulfur compounds (as S) 0.005% max
Thiophene To pass test (limit 1 ppm)
Water (H2O) 0.05% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR