Benzene, anhydrous ≥99.8%, DriSolv®, Supelco®

Supplier: MilliporeSigma
Danger


Packed in septum-sealed bottle.

BX0214-6 BX0214-7
EM-BX0214-6CS 935 USD
EM-BX0214-6 EM-BX0214-7
Benzene, anhydrous ≥99.8%, DriSolv®, Supelco®
Benzene

Packed under nitrogen. Flammable. Toxic.


Accessories information: Elbow adapter with septum cap (EM-692004-2) allows for the removal of solvent under nitrogen atmosphere. Replacement septum (EM-692004S-2), for use with elbow adapter, minimizes air and moisture intrusion once the primary septum is punctured. Drying tube (EM-692010-2) filled with t.h.e.* desiccant indicating silica gel removes solvent under a dry-air atmosphere. Large plug (EM-692012-2) is a replacement plug for bottle top.

Formula: C₆H₆
Boiling Pt: 80.1 °C (1013 hPa)
Melting Pt: 5.5 °C
Density: 0.878 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00003009
CAS Number: 71-43-2
EINECS: 200-753-7
UN: 1114
ADR: 3,II
REACH: 01-2119447106-44
Merck Index: 13,01066
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2c0146712ab33142aeeb3843b02381e6bda4028&vid=cae270bfe08ae6ab4b14d9e1775b89a82cfeec1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.8% min
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Residue after evaporation 5ppm max
Water (H2O) 30ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR