Trichloroacetic acid (TCA) 3% in toluene deblocking reagent

Supplier: MilliporeSigma

Synonyms: TCA, Trichloroethanoic acid, Aceto-Caustin, TCA

BI0837-4000
EM-BI0837-4000CS 2742 USD
EM-BI0837-4000
Trichloroacetic acid (TCA) 3% in toluene deblocking reagent
Trichloroacetic acid (TCA)

These deblocking reagents are mainly involved in DNA synthesis.


  • Infrared spectrum conforms to the standard
  • Corrosive and toxic
  • Clear liquid form

Formula: Cl₃CCOOH
MW: 163.39 g/mol
MDL Number: MFCD00004177
CAS Number: 76-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR