Capping B ACN/acetic anhydride/lutidine (50/20/30), Novabiochem® for DNA/RNA synthesis
Supplier: MilliporeSigma
BI0344-P4000
EM-BI0344-P4000CS 7278 USD
EM-BI0344-P4000
Capping B ACN/acetic anhydride/lutidine (50/20/30), Novabiochem® for DNA/RNA synthesis
Capping Reagent
These cappping reagents are involved in the formulations for oligonucleotide synthesis.
  • Soluble in water
  • Clear liquid form
  • Flammable, corrosive, and toxic effects
These are suitable for all kind of biosynthesis processes such as DNA, RNA and peptide synthesis.