Capping B 10% 1-Methylimidazole in tetrahydrofuran, Novabiochem® for DNA/RNA synthesis
Supplier: MilliporeSigma
BI0341-0450
EM-BI0341-0450CS 1078 USD
EM-BI0341-0450
Capping B 10% 1-Methylimidazole in tetrahydrofuran, Novabiochem® for DNA/RNA synthesis
Capping Reagent
These cappping reagents are involved in the formulations for oligonucleotide synthesis.
  • Soluble in water
  • Clear liquid form
  • Flammable, corrosive, and toxic effects
These are suitable for all kind of biosynthesis processes such as DNA, RNA and peptide synthesis.