5-(Ethylthio)-1H-tetrazole (ETT) 0.45 M in acetonitrile, Novabiochem® activator reagent, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 5-Ethyltio-1H-Tetrazole

BI0158-NP20
EM-BI0158-NP20EA 20203 USD
EM-BI0158-NP20
5-(Ethylthio)-1H-tetrazole (ETT) 0.45 M in acetonitrile, Novabiochem® activator reagent, Sigma-Aldrich®
5-(Ethylthio)-1H-tetrazole (ETT)

For DNA synthesis

Formula: C₃H₆N₄S
MW: 130.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00068733
CAS Number: 89797-68-2
UN: 2924
ADR: 3.8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR