L(+)-Ascorbic acid, GR ACS, Supelco®

Supplier: MilliporeSigma

AX1775-3 AX1775-3 AX1775-2 AX1775-5
CAAX1775-2EA 609.64 USD
CAAX1775-2 CAAX1775-3 EM-AX1775-3 EM-AX1775-2 EM-AX1775-5
L(+)-Ascorbic acid, GR ACS, Supelco®
L(+)-Ascorbic acid
Formula: C₆H₈O₆
Boiling Pt: 553 °C (1013 hPa)
Melting Pt: 190…192 °C
Density: 1.694 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064328
CAS Number: 50-81-7
EINECS: 200-066-2
REACH: exempt
Merck Index: 13,00837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min
Heavy metals (as Pb) 0.002% max
Iron (Fe) 0.001ppm max
Residue after ignition 0.1% max
Specific rotation [a]25/D (10%, water) +20.5-+21.5°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR