Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
1.09257.0500 1.88001.1045 1.88005.2500 1.88016.1000 1.09255.0500 1.09257.0500 1.88005.1000 1.88015.2500 1.88010.1045 1.88011.1045 1.88002.1045 1.88005.0500 1.88009.1000 1.88009.2500 1.88015.1000 1.88007.1002 1.88005.1003
CA1.09257.0500CS 5346.9 USD
CA1.09257.0500 EM-AX1697E-1 EM1.88001.1045 EM1.88005.2500 EM-1.88016.100 EM-AX1697A-1 EM-AX1697C-1 EM-AX1698E-6 EM1.09255.0500 EM1.09257.0500 EM1.88005.1000 EM1.88015.2500 EM-AX1697C-30 EM1.88010.1045 EM1.88011.1045 EM1.88002.1045 EM1.88005.0500 EM188009.1000 EM188009.2500 EM188015.1000 EM1.88007.1002 EM1.88005.1003
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Karl Fischer Titrants and Reagents
The Aquastar® range is designed to provide reagents for the determination of water by Karl Fischer titration of a wide range of substrates and by a variety of methodologies. These include reagents for one- or two-component systems; combined coulometric reagents; solvents for oils and fats; reagents and solvents for determination of water in ketones and aldehydes. Calibration standards are also available.
  • CombiTitrants and CombiSolvents are non toxic
  • Very accurate and reproducible results
  • Rapid and reliable
Merck have introduced into the range of Aquastar® Titrants, some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tags.