Isoamyl alcohol ≥98.5%, GR ACS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
AX1440-6
AX1440-3
EM-AX1440-6CS
1113.95
USD
EM-AX1440-6
EM-AX1440-3
Isoamyl alcohol ≥98.5%, GR ACS, Supelco®
Isoamyl alcohol
Meets ACS Specifications
Formula:
(CH₃)₂CHCH₂CH₂OH Boiling Pt: 130 °C (1013 hPa) Melting Pt: –117 °C Density: 0.812 g/cm³ (20 °C) Flash Pt: 42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002934 CAS Number: 123-51-3 EINECS: 204-633-5 UN: 1105 ADR: 3,III Merck Index: 13,05214 |
|
Specification Test Results
Assay (GC, corrected for water content) | 98.5% min |
Acids and esters (as amyl acetate) | 0.2% max |
Carbonyl compounds (as HCHO) | 0.1% max |
Color (APHA) | 10 max |
Form | Clear liquid |
Residue after evaporation | 0.003% max |
Titrable acid | 0.002meq/g max |
Water (H2O) | 0.5% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...