Aluminium sulfate octadecahydrate 98.0-102.0%, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Trade termforalum, Aluminum sulfate octadecahydrate, Aluminum(III) sulfate octadecahydrate, Aluminum(III) sulphate octadecahydrate, Aluminum sulphate octadecahydrate

AX0735-3
EM-AX0735-1EA 303.53 USD
EM-AX0735-1 EM-AX0735-3
Aluminium sulfate octadecahydrate 98.0-102.0%, GR ACS, Supelco®
Aluminium sulphate octadecahydrate

Meets ACS Specifications

Formula: Al₂(SO₄)₃·18H₂O
MW: 666.43 g/mol
Melting Pt: 86
Density: 1.69 g/cm³ (20 °C)
MDL Number: MFCD00149136
CAS Number: 7784-31-8
EINECS: 233-135-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dace68759944aa1c6c0f56a372ea396f004e261&vid=e63de3c58650a97622a605c237aa52402ff157e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (complexometric) 98.0-102.0%
Calcium (Ca) 0.01% max
Chloride (Cl) 0.005% max
Color White
Form Crystals
Heavy metals (as Pb) 0.001% max
Insoluble matter 0.01% max
Iron (Fe) 0.002% max
Magnesium (Mg) 0.002% max
Potassium (K) 0.005% max
Sodium (Na) 0.02% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR