Alcohol ≥82-86%, Reagent Grade (denatured with methanol and isopropanol), Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Reagent alcohol

AX0442-3
EM-AX0442-3CS 1099 USD
EM-AX0442-3
Alcohol ≥82-86%, Reagent Grade (denatured with methanol and isopropanol), Supelco®
Alcohol
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR