Acetonitrile ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
AX0156-1
AX0156-1
AX0156-6
CAAX0156-1CS
5259.99
USD
CAAX0156-1
EM-AX0156-1
EM-AX0156-6
Acetonitrile ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Acetonitrile
Clear liquid. For use in HPLC and spectrophotometry. Filtered through a 0.2µm element for purity. UV cutoff at 190nm.
Formula:
H₃CCN MW: 41.05 g/mol Boiling Pt: 81.6 Melting Pt: -45.7 Density: 0.782 g/cm³ (20 °C) Flash Pt: 2 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3 II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Specification Test Results
Assay (GC) | 99.9% min |
Color (APHA) | 10APHA max |
Filtered through 0.2 µm filter | To pass test |
Fluorescence as quinine at 254 nm | 1ppb max |
Fluorescence as quinine at 365 nm | 1ppb max |
Form | Clear liquid |
Gradient at 210 nm | 2mAU max |
Gradient at 254 nm | 0.3mAU max |
Identity (IR-spectrum) | Conforms |
Metals, suitable for LC/MS | To pass test |
MS purity, ESI(+), largest response as reserpine | 50ppb max |
MS purity, ESI(-), largest response, p-nitrophenol | 50ppb max |
Residue after evaporation | 1ppm max |
Titrable acid | 1µeq/g max |
Titrable base | 0.5µeq/g max |
UV Abs. at 200 nm | 0.1AU max |
UV Abs. at 210 nm | 0.04AU max |
UV Abs. at 220 nm | 0.02AU max |
UV Abs. at 230 nm | 0.01AU max |
UV Abs. at 240 nm | 0.01AU max |
UV Abs. at 250 nm | 0.01AU max |
UV Abs. at 270 nm | 0.01AU max |
UV Abs. at 400-280 nm | 0.005AU max |
UV Cut-off | 190nm max |
Water (H2O) | 0.02% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...