Acetonitrile ≥99.9%, OmniSolv®, non-UV for gas chromatography, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
AX0155-1
EM-AX0155-1CS
2137
USD
EM-AX0155-1
Acetonitrile ≥99.9%, OmniSolv®, non-UV for gas chromatography, Supelco®
Acetonitrile
For GC and residue analysis. Not optically transparent in the UV region. Certificate of Analysis on label.
Formula:
H₃CCN MW: 41.05 g/mol Boiling Pt: 81.6 °C (1013 hPa) Melting Pt: –45.7 °C Density: 0.782 g/cm³ (20 °C) Flash Pt: 2 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
Specification Test Results
Assay (GC) | 99.9% min |
Capillary ECD responsive substances (as PCNB) | 2ng/L max |
Color (APHA) | 10 max |
Filtered through 0.2 µm filter | To pass test |
Form | Clear liquid |
Infrared spectrum | Conforms to standard |
Residue after evaporation | 1ppm max |
Titrable acid | 8.3µeq/g max |
Titrable base | 0.6µeq/g max |
Water (H2O) | 0.10% max |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...