Acetonitrile, anhydrous, Supelco®

Supplier: MilliporeSigma
Danger

AX0146-6
EM-AX0146-6CS 1139 USD
EM-AX0146-6
Acetonitrile, anhydrous, Supelco®
Acetonitrile

For nonaqueous titrations.

Formula: H₃CCN
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10 max
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 0.001% max
Water (H2O) 0.01% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR