Acetonitrile ≥99.8% for HPLC, Supelco®

Supplier: MilliporeSigma
Danger

AX0145-1 AX0145-NP20 AX0145P-6 AX0145P-1 AX0145-1
CAAX0145-1CS 3515.22 USD
CAAX0145-1 EM-AX0145-39 EM-AX0145-31 EM-AX0145-NP20 EM-AX0145P-6 EM-AX0145P-1 EM-AX0145-1
Acetonitrile ≥99.8% for HPLC, Supelco®
Acetonitrile

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: H₃CCN
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid, free from particulates
Assay (GC) 99.8% min
Color (APHA) 10 max
Residue after evaporation 1ppm max
Filtered through 0.2 µm filter To pass test
Gradient at 254 nm 0.005AU max
Infrared spectrum Conforms to standard
Titrable acid 0.008meq/g max
Titrable base 0.0006meq/g max
UV Abs. at 190 nm 1.00AU max
UV Abs. at 220 nm 0.05AU max
UV Abs. at 254 - 400 nm 0.01AU max
UV Cut-off 190nm max
Water (H2O) 0.10% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR