Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

AX0142-6 AX0142P-1 AX0142-6 AX0142-NP20 AX0142P-1 AX0142-1
CAAX0142-6CS 1514.66 USD
CAAX0142-6 CAAX0142P-1 EM-AX0142-6 EM-AX0142-31 EM-AX0142-NP20 EM-AX0142P-1 EM-AX0142-1
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Acetonitrile

For spectrophotometry and chromatography. UV cutoff 189 nm. Gradient controlled to remove lipophilic interferences at 254 and 210 nm. Certificate of lot analysis on label.

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 300ppt max
Form Clear liquid
Gradient at 210 nm 0.002AU max
Gradient at 254 nm 0.0003AU max
Identity (IR-spectrum) Conforms
Residue after evaporation 1ppm max
Titrable acid 8.0µeq/g max
Titrable base 0.16µeq/g max
UV Abs. at 189 nm 1.00AU max
UV Abs. at 195 nm 0.15AU max
UV Abs. at 210 nm 0.03AU max
UV Abs. at 230 nm 0.005AU max
UV Abs. at 250 nm 0.005AU max
UV Abs. at 270 nm 0.005AU max
UV Cut-off 189nm max
Water (H2O) 0.02% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR