Acetone ≥99% (by GC) for histology, Supelco®

Supplier: MilliporeSigma
Danger

AX0125-4 AX0125-20 AX0125-3 AX0125-4 AX0125-6 AX0125-20
CAAX0125-4CS 3541.4 USD
CAAX0125-4 EM-AX0125-20 EM-AX0125-3 EM-AX0125-4 EM-AX0125-6 TXAX0125-20BSH
Acetone ≥99% (by GC) for histology, Supelco®
Acetone

Most lipids are removed from tissues with the agent.

Formula: CH₃COCH₃
Boiling Pt: 56 °C/760 mmHg (lit.)
Melting Pt: –94 °C (lit.)
Density: 0.791 g/ml (25°C) (lit.)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99% min
Color (APHA) 10 max
Form Clear liquid
Identification To pass test
Infrared spectrum Conforms to standard
Readily oxidizable substances To pass test
Residue after evaporation 0.001% max
Solubility To pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR