Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

AX0120-3 AX0120-2 AX0120-8 AX0120-6 AX0120-2 AX0120-3 AX0120P-4 AX0120-30 AX0120-9 AX0120-8 AX0120-20
CAAX0120-3EA 2746.91 USD
CAAX0120-3 CAAX0120-2 CAAX0120-8 EM-AX0120-6 EM-AX0120-2 EM-AX0120-3 EMD-AX0120P-4 EM-AX0120-30 EM-AX0120-9 EMD-AX0120-8 EM-AX0120-20 EM-AX0120T-4
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Acetone

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: CH₃COCH₃
Boiling Pt: 56 °C/760 mmHg (lit.)
Melting Pt: –94 °C (lit.)
Density: 0.791 g/ml (25°C) (lit.)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC, corrected for water content) 99.5% min
Aldehyde (as HCHO) 0.002% max
Color (APHA) 10 max
Form Clear liquid
Isopropyl alcohol 0.05% max
Methanol 0.05% max
Residue after evaporation 0.001% max
Solubility in water To pass test
Substances reducing permanganate To pass test
Titrable acid 0.0003meq/g max
Titrable base 0.0006meq/g max
Water (H2O) 0.5% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR