TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), OmniPur®, Millipore®

Supplier: MilliporeSigma
Danger

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

9400-100ML 9410-1L 9440-4L 9460-20L
EM-9400EA 76.1 USD
EM-9400 EM-9410 EM-9440 EM-9460
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), OmniPur®, Millipore®
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Melting Pt: 6
Flash Pt: 251 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9 III
Merck Index: 13,06793

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR