TE Buffer, Concentrate, OmniPur*. 100X.
Supplier: MilliporeSigma
8910-1L
EM-8910EA 432.76 USD
EM-8910
TE Buffer, Concentrate, OmniPur*. 100X.
Buffers Electrophoresis Buffers Running Buffers
Colorless, hazefree liquid
TRIS base 1.0M, Na2EDTA 0.10M (in 100X concentrate). For molecular biology. DNase-, RNase-, and protease-free.